Assay Details
| Gene Symbol: Cdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039154.2 | NP_001034243.1 | 7 - 8 | 1863 | 65 | |
| NM_001285913.1 | NP_001272842.1 | 7 - 8 | 1863 | 65 | ||
| NM_001285914.1 | NP_001272843.1 | 7 - 8 | 1863 | 65 | ||
| NM_007667.3 | NP_031693.2 | 7 - 8 | 1863 | 65 | ||
| GenBank mRNA | AK044046.1 | - | 7 - 8 | 1821 | 65 | |
| AK047589.1 | - | 5 - 6 | 794 | 65 | ||
| AK083092.1 | - | 7 - 8 | 1830 | 65 | ||
| AK086711.1 | - | 7 - 8 | 1832 | 65 | ||
| AK139334.1 | - | 7 - 8 | 1819 | 65 | ||
| BC057581.1 | - | 7 - 8 | 1768 | 6416918 | 65 | |
| BC060200.1 | - | 7 - 8 | 1711 | 65 | ||
| X95600.1 | - | 7 - 8 | 1503 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh8
Genomic Map