Assay Details
Gene Symbol: Efcab5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176965.3 | NP_795939.2 | 15 - 16 | 3698 | 66 | |
GenBank mRNA | AK029790.1 | - | 13 - 14 | 3285 | 66 | |
AK035698.1 | - | 11 - 12 | 2532 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
