Assay Details
Gene Symbol: Efcab5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176965.3 | NP_795939.2 | 8 - 9 | 2348 | 147 | |
GenBank mRNA | AK029790.1 | - | 6 - 7 | 1935 | 147 | |
AK035698.1 | - | 4 - 5 | 1182 | 147 | ||
BC127154.1 | - | 6 - 7 | 1890 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
