Assay Details
| Gene Symbol: Cdh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009866.4 | NP_033996.4 | 10 - 11 | 2034 | 58 | |
| GenBank mRNA | AK028271.1 | - | 6 - 7 | 921 | 58 | |
| AK049652.1 | - | 10 - 11 | 2000 | 58 | ||
| BC046314.1 | - | 10 - 11 | 1934 | 6312939 | 58 | |
| D21253.1 | - | 9 - 10 | 1797 | 58 | ||
| D31963.1 | - | 10 - 11 | 1979 | 58 | ||
| X77557.1 | - | 10 - 11 | 1943 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh11
Genomic Map