Assay Details
| Gene Symbol: Cdh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009866.4 | NP_033996.4 | 7 - 8 | 1509 | 77 | |
| GenBank mRNA | AK028271.1 | - | 3 - 4 | 396 | 77 | |
| AK049652.1 | - | 7 - 8 | 1475 | 77 | ||
| BC046314.1 | - | 7 - 8 | 1409 | 6312939 | 77 | |
| D21253.1 | - | 6 - 7 | 1272 | 77 | ||
| D31963.1 | - | 7 - 8 | 1454 | 77 | ||
| X77557.1 | - | 7 - 8 | 1418 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh11
Genomic Map