Assay Details
| Gene Symbol: Cdh11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009866.4 | NP_033996.4 | 8 - 9 | 1757 | 142 | |
| GenBank mRNA | AK028271.1 | - | 4 - 5 | 644 | 142 | |
| AK049652.1 | - | 8 - 9 | 1723 | 142 | ||
| BC046314.1 | - | 8 - 9 | 1657 | 6312939 | 142 | |
| D21253.1 | - | 7 - 8 | 1520 | 142 | ||
| D31963.1 | - | 8 - 9 | 1702 | 142 | ||
| X77557.1 | - | 8 - 9 | 1666 | 142 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh11
Genomic Map