Assay Details
| Gene Symbol: P2rx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033321.3 | NP_201578.2 | 11 - 12 | 1198 | 73 | |
| GenBank mRNA | AF333331.1 | - | 11 - 12 | 1204 | 73 | |
| AK134662.1 | - | 11 - 12 | 1158 | 73 | ||
| BC138189.1 | - | 11 - 12 | 1198 | 8861208 | 73 | |
| BC145181.1 | - | 11 - 12 | 1198 | 9053732 | 73 | |
| JX570808.1 | - | 10 - 11 | 1461 | 73 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

P2rx5
Genomic Map