Assay Details
Gene Symbol: Cdk17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146239.2 | NP_666351.2 | 2 - 3 | 470 | 62 | |
GenBank mRNA | AK078325.1 | - | 2 - 3 | 471 | 62 | |
AK203943.1 | - | 1 - 2 | 156 | 62 | ||
AK205070.1 | - | 1 - 2 | 156 | 62 | ||
BC064815.1 | - | 2 - 3 | 455 | 6516981 | 62 | |
CD350146.1 | - | 1 - 2 | 100 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
