Assay Details
| Gene Symbol: Rbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282000.1 | NP_001268929.1 | 11 - 12 | 1676 | 123 | |
| NM_001282001.1 | NP_001268930.1 | 12 - 13 | 1775 | 123 | ||
| NM_011250.4 | NP_035380.3 | 12 - 13 | 1805 | 123 | ||
| GenBank mRNA | AK160027.1 | - | 12 - 13 | 1752 | 123 | |
| BC020528.1 | - | 12 - 13 | 1753 | 4240592 | 123 | |
| U36799.1 | - | 12 - 13 | 1675 | 123 | ||
| U47333.1 | - | 11 - 12 | 1618 | 123 | ||
| U50850.1 | - | 12 - 13 | 1713 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbl2
Genomic Map