Assay Details
| Gene Symbol: Pecam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK169431.1 | - | 13 - 14 | 2034 | 74 | |
| L06039.1 | - | 14 - 15 | 2224 | 74 | ||
| S79861.1 | - | 6 - 7 | 299 | 74 | ||
More Information
Set Membership: |
|

Pecam1
Genomic Map