Assay Details
Gene Symbol: Pecam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK037551.1 | - | 2 - 3 | 646 | 92 | |
AK084527.1 | - | 1 - 2 | 235 | 92 | ||
AK169431.1 | - | 1 - 2 | 238 | 92 | ||
BC008519.1 | - | 1 - 2 | 240 | 3592726 | 92 | |
BC085502.1 | - | 1 - 2 | 42 | 5720681 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK037551.1, AK169431.1, BC008519.1, BC085502.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
