Assay Details
| Gene Symbol: Tcam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029467.3 | NP_083743.2 | 7 - 8 | 1659 | 68 | |
| GenBank mRNA | AK076581.1 | - | 7 - 8 | 1745 | 68 | |
| BC050746.1 | - | 1583 | 6741794 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013475 | BC050746.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2013475 | NM_029467.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 32600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625378 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 74902 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 903933 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625334 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 960962 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 818027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 155698 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172178 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151655 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013475 | BC050746.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2013475 | NM_029467.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100638 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100649 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100647 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100650 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100645 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28100652 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151655 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172178 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07479332 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07479356 | - | - |
| Whole Mouse Genome G4122A | A_51_P226199 | BC050746.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P226199 | AK016263.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P226199 | AK076581.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P226199 | NM_029467.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P293391 | BC050746.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P293391 | AK076581.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P293391 | NM_029467.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P123692 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172178 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151655 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09079970 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09079967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100579 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100719 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100569 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100606 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100721 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100609 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100571 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28100578 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09430378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09334269 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100638 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09055697 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09709769 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09334266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09585783 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100670 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09432145 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09709766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09079965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09102980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100664 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100660 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100609 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100652 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100642 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09460245 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09373235 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09687811 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09379997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09379981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100640 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28100654 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10382088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382087 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382095 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382089 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382090 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382084 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382092 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382093 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10382094 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421540_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435410_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10382083 | - | - |
| Mouse Genome 430 2.0 Array | 1421540_at | - | - |
| Mouse Genome 430 2.0 Array | 1435410_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1435410_at | - | - |
| Mouse Genome 430 2.0 Array | 1421540_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75870_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75870_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100016829.mm.1 | NM_029467.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100016829.mm.1 | NM_029467.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100016829.mm.1 | TC1100001780.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100016830.mm.1 | NM_029467.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100016830.mm.1 | NM_029467.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100016830.mm.1 | TC1100001780.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100016832.mm.1 | TC1100001780.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1421540_PM_at | - | - |
| Mouse Expression Set 430 | 1421540_at | - | - |
| Mouse Expression Set 430 | 1435410_at | - | - |
| Mouse Expression Set 430 | 1435410_PM_at | - | - |
| Murine Genome U74v2 Set | 169570_PM_f_at | - | - |
| Murine Genome U74v2 Set | 169570_f_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421540_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421540_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421540_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421540_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435410_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435410_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1170433 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6350301 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1170433 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006350301 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001170433 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2648952 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1258927 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2648952 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1258927 | - | - |
| mouse-6 v1.1 (Illumina) | scl40796.9.1_58-S | - | - |
| mouse-6 v1.1 (Illumina) | scl0075870.2_314-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6350301 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1170433 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40796.9.1_58-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0075870.2_314-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1170433 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6350301 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1258927 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2648949 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2912865 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2648949 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1258927 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2912865 | - | - |

Tcam1
Genomic Map