Assay Details
| Gene Symbol: Atp2c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026922.1 | NP_081198.1 | 11 - 12 | 1135 | 64 | |
| GenBank mRNA | AK137687.1 | - | 11 - 12 | 1136 | 64 | |
| AK220351.1 | - | 9 - 10 | 720 | 64 | ||
| BC042772.1 | - | 2 - 3 | 153 | 64 | ||
| EF688288.1 | - | 11 - 12 | 979 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971931 | EF688288.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971931 | BC026636.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971931 | BC037514.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971931 | AK220351.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971931 | BC042772.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 92477 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700475 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283774 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84772 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525425 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 180233 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 853211 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 751441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 573313 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654061 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 650707 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 368617 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396224 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 7544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669026 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631042 | AK144570.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 631042 | BC029710.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 862066 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 710959 | AK014816.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 204458 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 773438 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 594336 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 653199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39564 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 933600 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15427 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 581848 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 496434 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 277841 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 236490 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 596391 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 361181 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 357795 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 278433 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195609 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 148926 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73422 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 136135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 72952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 19606 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P130416 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159207 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971931 | EF688288.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971931 | BC026636.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971931 | BC037514.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971931 | AK220351.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971931 | BC042772.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06662353 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06662410 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06662319 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P130416 | - | - |
| Whole Mouse Genome G4122A | A_51_P189121 | BC026636.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189121 | BC037514.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189121 | BC042772.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189121 | NM_026922.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P189121 | AK007419.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135971 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06662319 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P130416 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07000573 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215498 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06681084 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07000574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215776 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06784703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06784711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07025105 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215538 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07025110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07060810 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215562 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07003627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215502 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07060813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215564 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06949921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215547 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215534 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215556 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07183805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215543 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26215536 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10575950 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575935 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575928 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575931 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575939 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575936 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575953 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575951 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575932 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575944 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575937 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575954 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575947 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575938 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575930 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575941 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575948 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575952 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575949 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575942 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575933 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575943 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575927 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575934 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575945 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575940 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575946 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10575929 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452326_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10575926 | - | - |
| Mouse Genome 430 2.0 Array | 1452326_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452326_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69047_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69047_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800011622.mm.1 | NM_026922.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011622.mm.1 | NM_026922.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011622.mm.1 | TC0800001426.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800011623.mm.1 | NM_026922.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011623.mm.1 | NM_026922.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800011623.mm.1 | TC0800001426.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1452326_PM_at | - | - |
| Mouse Expression Set 430 | 1452326_at | - | - |
| Murine Genome U74v2 Set | 165918_at | - | - |
| Murine Genome U74v2 Set | 165918_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452326_at | - | - |
| Mouse Genome 430A 2.0 Array | 1452326_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452326_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1452326_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 921510 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1257530 | - | - |

Atp2c2
Genomic Map