Assay Details
| Gene Symbol: Necab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_054095.2 | NP_473436.1 | 4 - 5 | 701 | 76 | |
| GenBank mRNA | AF411253.1 | - | 4 - 5 | 695 | 76 | |
| AK035168.1 | - | 2 - 3 | 76 | 76 | ||
| AK082727.1 | - | 4 - 5 | 311 | 76 | ||
| BC107384.1 | - | 5 - 6 | 296 | 76 | ||
| BC107385.1 | - | 5 - 6 | 260 | 76 | ||
| BC115857.1 | - | 5 - 6 | 233 | 40057629 | 76 | |
| BC115963.1 | - | 5 - 6 | 221 | 40057628 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC115857.1, BC115963.1 transcripts.
Set Membership: |
|

Necab2
Genomic Map