Assay Details
| Gene Symbol: Ftsj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025310.2 | NP_079586.1 | 4 - 5 | 291 | 84 | |
| GenBank mRNA | AK004996.2 | - | 4 - 5 | 291 | 84 | |
| AK031514.1 | - | 4 - 5 | 278 | 84 | ||
| AK145490.1 | - | 4 - 5 | 316 | 84 | ||
| BC012281.1 | - | 4 - 5 | 288 | 4012961 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ftsj3
Genomic Map