Assay Details
| Gene Symbol: Fam195b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033231.2 | NP_001028403.1 | 4 - 5 | 344 | 79 | |
| GenBank mRNA | AK147780.1 | - | 4 - 5 | 322 | 79 | |
| BC002190.1 | - | 3 - 4 | 276 | 79 | ||
| BC003940.1 | - | 3 - 4 | 276 | 79 | ||
| BC025536.1 | - | 4 - 5 | 317 | 79 | ||
| BC052229.1 | - | 3 - 4 | 446 | 79 | ||
| BI987638.1 | - | 4 - 5 | 344 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185236 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185235 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185234 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185233 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185237 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985077 | BC002190.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985077 | BC003940.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985077 | BC025536.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985077 | BC052229.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985077 | BI987638.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985079 | BC002190.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985079 | BC003940.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985079 | BC025536.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985079 | BC052229.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1985079 | AK210372.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891367 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909185 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 131886 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59826 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 165909 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128065 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985077 | BC002190.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985077 | BC003940.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985077 | BC025536.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985077 | BC052229.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985077 | BI987638.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985079 | BC002190.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985079 | BC003940.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985079 | BC025536.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985079 | BC052229.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1985079 | AK210372.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185224 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185245 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185266 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185279 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185241 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185283 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185238 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185237 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185272 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185233 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28185281 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07516656 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128065 | - | - |
| Whole Mouse Genome G4122A | A_51_P176138 | BC002190.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176138 | BC003940.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176138 | BC025536.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176138 | BC052229.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P176138 | NM_001033231.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128065 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185235 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185237 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185236 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185234 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28185233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185132 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185235 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185113 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185282 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185200 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185205 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185279 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28185275 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185229 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185197 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185204 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09733960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185191 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185232 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185223 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185209 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09483532 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185233 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09299104 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185226 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09383108 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185178 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185195 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185224 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09299114 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185205 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185207 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09733961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185182 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09385813 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185217 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185212 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185208 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185185 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P28185216 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10393817 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393816 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10393815 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1460690_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10393814 | - | - |
| Mouse Genome 430 2.0 Array | 1460690_at | - | - |
| Mouse Genome 430 2.0 Array | 1460690_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192173_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 192173_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100040179.mm.1 | NM_001033231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040179.mm.1 | NM_001033231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040179.mm.1 | TC1100004212.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100040183.mm.1 | NM_001033231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040183.mm.1 | NM_001033231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040183.mm.1 | TC1100004212.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100040185.mm.1 | NM_001033231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040185.mm.1 | NM_001033231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040185.mm.1 | TC1100004212.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100040180.mm.1 | NM_001033231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040180.mm.1 | NM_001033231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040180.mm.1 | TC1100004212.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100040181.mm.1 | NM_001033231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040181.mm.1 | NM_001033231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040181.mm.1 | TC1100004212.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100040184.mm.1 | NM_001033231.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040184.mm.1 | NM_001033231.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100040184.mm.1 | TC1100004212.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1460690_PM_at | - | - |
| Mouse Expression Set 430 | 1460690_at | - | - |
| Murine Genome U74v2 Set | 104559_PM_at | - | - |
| Murine Genome U74v2 Set | 104559_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460690_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1460690_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460690_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1460690_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232298 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232298 | - | - |

Fam195b
Genomic Map