Assay Details
Gene Symbol: Lrrc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153545.2 | NP_705773.2 | 3 - 4 | 421 | 86 | |
GenBank mRNA | AK081566.1 | - | 3 - 4 | 424 | 86 | |
AK153361.1 | - | 3 - 4 | 422 | 86 | ||
BC023296.1 | - | 3 - 4 | 401 | 3662961 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
