Assay Details
| Gene Symbol: P4hb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011032.2 | NP_035162.1 | 7 - 8 | 1196 | 73 | |
| GenBank mRNA | AK141304.1 | - | 7 - 8 | 1133 | 73 | |
| AK146542.1 | - | 7 - 8 | 1134 | 73 | ||
| AK149605.1 | - | 7 - 8 | 1135 | 73 | ||
| AK149965.1 | - | 7 - 8 | 1134 | 73 | ||
| AK150002.1 | - | 7 - 8 | 1160 | 73 | ||
| AK150422.1 | - | 7 - 8 | 1134 | 73 | ||
| AK150592.1 | - | 7 - 8 | 1134 | 73 | ||
| AK150755.1 | - | 7 - 8 | 1120 | 73 | ||
| AK150805.1 | - | 7 - 8 | 1134 | 73 | ||
| AK151351.1 | - | 7 - 8 | 1128 | 73 | ||
| AK151546.1 | - | 7 - 8 | 1134 | 73 | ||
| AK152141.1 | - | 7 - 8 | 1134 | 73 | ||
| AK152217.1 | - | 7 - 8 | 1134 | 73 | ||
| AK152791.1 | - | 7 - 8 | 1134 | 73 | ||
| AK152834.1 | - | 7 - 8 | 1134 | 73 | ||
| AK153365.1 | - | 7 - 8 | 1134 | 73 | ||
| AK159606.1 | - | 7 - 8 | 1134 | 73 | ||
| AK159729.1 | - | 7 - 8 | 1134 | 73 | ||
| AK159965.1 | - | 7 - 8 | 1134 | 73 | ||
| AK161537.1 | - | 7 - 8 | 1134 | 73 | ||
| AK168330.1 | - | 7 - 8 | 1132 | 73 | ||
| AK168336.1 | - | 7 - 8 | 1116 | 73 | ||
| AK168612.1 | - | 7 - 8 | 1134 | 73 | ||
| AK168893.1 | - | 7 - 8 | 1134 | 73 | ||
| AK169265.1 | - | 7 - 8 | 1131 | 73 | ||
| AK170603.1 | - | 7 - 8 | 1131 | 73 | ||
| BC008549.1 | - | 7 - 8 | 1122 | 3591404 | 73 | |
| BC093512.1 | - | 7 - 8 | 1113 | 5124376 | 73 | |
| J05185.1 | - | 7 - 8 | 1196 | 73 | ||
| X06453.1 | - | 7 - 8 | 1136 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

P4hb
Genomic Map