| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800022580.mm.1 |
NM_007916.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022580.mm.1 |
NM_007916.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022580.mm.1 |
TC0800002934.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800022578.mm.1 |
NM_007916.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022578.mm.1 |
NM_007916.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022578.mm.1 |
TC0800002934.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800022579.mm.1 |
NM_007916.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022579.mm.1 |
NM_007916.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800022579.mm.1 |
TC0800002934.mm.1 |
MTA TC |