Assay Details
| Gene Symbol: Ddx19a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007916.2 | NP_031942.2 | 2 - 3 | 213 | 85 | |
| GenBank mRNA | AK049467.1 | - | 2 - 3 | 184 | 85 | |
| AK146548.1 | - | 2 - 3 | 174 | 85 | ||
| BC011270.1 | - | 2 - 3 | 185 | 3674608 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx19a
Genomic Map