Assay Details
Gene Symbol: Fanca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016925.3 | NP_058621.2 | 27 - 28 | 2609 | 64 | |
GenBank mRNA | AF178934.1 | - | 27 - 28 | 2605 | 64 | |
AF208116.1 | - | 27 - 28 | 2611 | 64 | ||
AF208121.1 | - | 2 - 3 | 256 | 64 | ||
BC150705.1 | - | 27 - 28 | 2607 | 9087616 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
