Assay Details
| Gene Symbol: Pdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198308.1 | NP_938050.1 | 11 - 12 | 1602 | 99 | |
| GenBank mRNA | AK029575.1 | - | 11 - 12 | 1693 | 99 | |
| AK078515.1 | - | 5 - 6 | 693 | 99 | ||
| AK129472.1 | - | 11 - 12 | 1706 | 99 | ||
| AY223867.1 | - | 11 - 12 | 1602 | 99 | ||
| BC120745.1 | - | 10 - 11 | 1532 | 40129668 | 99 | |
| BC125425.1 | - | 10 - 11 | 1532 | 40129940 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdpr
Genomic Map