Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800010930.mm.1 |
NM_198308.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010930.mm.1 |
NM_198308.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010930.mm.1 |
TC0800001328.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010929.mm.1 |
NM_198308.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010929.mm.1 |
NM_198308.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010929.mm.1 |
TC0800001328.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010932.mm.1 |
NM_198308.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010932.mm.1 |
NM_198308.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010932.mm.1 |
TC0800001328.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800010931.mm.1 |
NM_198308.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010931.mm.1 |
NM_198308.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800010931.mm.1 |
TC0800001328.mm.1 |
MTA TC |