Assay Details
| Gene Symbol: Pdpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198308.1 | NP_938050.1 | 7 - 8 | 976 | 109 | |
| GenBank mRNA | AK029575.1 | - | 7 - 8 | 1067 | 109 | |
| AK078515.1 | - | 1 - 2 | 67 | 110 | ||
| AK129472.1 | - | 7 - 8 | 1080 | 109 | ||
| AY223867.1 | - | 7 - 8 | 976 | 109 | ||
| BC120745.1 | - | 6 - 7 | 906 | 40129668 | 109 | |
| BC125425.1 | - | 6 - 7 | 906 | 40129940 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pdpr
Genomic Map