Assay Details
Gene Symbol: Ednra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010332.2 | NP_034462.1 | 4 - 5 | 1157 | 80 | |
GenBank mRNA | AF039892.1 | - | 3 - 4 | 452 | 80 | |
AK017486.1 | - | 4 - 5 | 1152 | 80 | ||
AK042211.1 | - | 4 - 5 | 1171 | 80 | ||
AK043210.1 | - | 4 - 5 | 1054 | 80 | ||
AK135555.1 | - | 4 - 5 | 1051 | 80 | ||
BC008277.1 | - | 4 - 5 | 1141 | 2812426 | 80 | |
L20340.1 | - | 1 - 2 | 154 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
