Assay Details
Gene Symbol: Ednra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010332.2 | NP_034462.1 | 5 - 6 | 1310 | 70 | |
GenBank mRNA | AF039892.1 | - | 4 - 5 | 605 | 70 | |
AK043210.1 | - | 5 - 6 | 1207 | 70 | ||
AK135555.1 | - | 5 - 6 | 1204 | 70 | ||
BC008277.1 | - | 5 - 6 | 1294 | 2812426 | 70 | |
L20340.1 | - | 2 - 3 | 307 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
