Assay Details
Gene Symbol: Isx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027837.3 | NP_082113.2 | 3 - 4 | 950 | 60 | |
GenBank mRNA | AB219124.1 | - | 3 - 4 | 483 | 60 | |
AK018615.1 | - | 3 - 4 | 951 | 60 | ||
BC128290.1 | - | 3 - 4 | 584 | 40102443 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252988 | NM_027837.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252988 | AK018615.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252988 | NM_027837.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P252988 | AK018615.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900899 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827507 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 59523 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766373 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 667947 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93865 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934245 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285762 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 827416 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834772 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 187912 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415767 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234323 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342801 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26774 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 87415 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P152066 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160011 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252988 | NM_027837.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252988 | AK018615.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252988 | NM_027837.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P252988 | AK018615.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160011 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P152066 | - | - |
Whole Mouse Genome G4122A | A_51_P252988 | NM_027837.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P252988 | AK018615.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P252988 | NM_027837.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P252988 | AK018615.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P152066 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970748 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970739 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970762 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970858 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970732 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970772 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970734 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970948 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970851 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970932 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970866 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970926 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970924 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970911 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970750 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970893 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970721 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25970843 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970699 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07327191 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06712813 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970704 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970904 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970753 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970788 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970764 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970759 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970731 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970811 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07004842 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970809 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970791 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06932300 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07166738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06932297 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06932314 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06932294 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970729 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970717 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970756 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970800 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970918 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970908 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970812 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07315601 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06772648 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970706 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06747436 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970709 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07004845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970950 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970793 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970771 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06956496 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06772639 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970777 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970727 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970912 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970748 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970786 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07332825 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970903 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06932305 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970802 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970746 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970799 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970779 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970707 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06669276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970785 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970944 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970913 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07327189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07261567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970790 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970784 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970696 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970906 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06747445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970708 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06932312 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970718 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970935 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06956493 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06992957 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07424407 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970794 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07327193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970798 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06932289 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25970942 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06956498 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10572867 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10572868 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10572869 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10572866 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1431922_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10572865 | - | - |
Mouse Genome 430 2.0 Array | 1431922_at | - | - |
Mouse Genome 430 2.0 Array | 1431922_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71597_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71597_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800006209.mm.1 | NM_027837.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800006209.mm.1 | NM_027837.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800006209.mm.1 | TC0800000842.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800006208.mm.1 | NM_027837.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800006208.mm.1 | NM_027837.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800006208.mm.1 | TC0800000842.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800006210.mm.1 | NM_027837.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800006210.mm.1 | NM_027837.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800006210.mm.1 | TC0800000842.mm.1 | MTA TC |
Mouse Expression Set 430 | 1431922_at | - | - |
Mouse Expression Set 430 | 1431922_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431922_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1431922_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 368751 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240449 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1240449 | - | - |
