Assay Details
| Gene Symbol: Tnnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009394.2 | NP_033420.1 | 3 - 4 | 270 | 97 | |
| GenBank mRNA | AK133548.1 | - | 3 - 4 | 266 | 97 | |
| BC024390.1 | - | 3 - 4 | 269 | 5026329 | 97 | |
| CB566378.1 | - | 305 | 97 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Tnnc2
Genomic Map