Assay Details
| Gene Symbol: Tnnc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009394.2 | NP_033420.1 | 5 - 6 | 516 | 91 | |
| GenBank mRNA | AK133548.1 | - | 5 - 6 | 512 | 91 | |
| BC024390.1 | - | 5 - 6 | 515 | 5026329 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819733 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819735 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819734 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819737 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819736 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353232 | NM_009394.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353232 | BC024390.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353232 | NM_009394.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353232 | BC024390.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 884310 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145324 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353232 | NM_009394.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353232 | BC024390.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353232 | NM_009394.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353232 | BC024390.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353232 | NM_009394.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353232 | BC024390.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353232 | NM_009394.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353232 | BC024390.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819728 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819736 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819731 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819716 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819720 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819733 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21819691 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145324 | - | - |
| Whole Mouse Genome G4122A | A_51_P353232 | NM_009394.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353232 | BC024390.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353232 | NM_009394.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353232 | BC024390.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138214 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145324 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02111460 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02111444 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01420734 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01420738 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01420725 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02111450 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819733 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819736 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819735 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819734 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21819737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819760 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819758 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819752 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02111423 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819721 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819717 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819679 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819720 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02111441 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02111421 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02111460 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02111443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819712 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02099956 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02111427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819716 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819714 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819705 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819692 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02257704 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02111454 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819707 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819696 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02099955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819687 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21819680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02111457 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10489551 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489548 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489552 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489549 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489550 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489547 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489546 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417464_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10478572 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10489545 | - | - |
| Mouse Genome 430 2.0 Array | 1417464_at | - | - |
| Mouse Genome 430 2.0 Array | 1417464_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21925_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21925_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200043735.mm.1 | NM_009394.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043735.mm.1 | NM_009394.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200043735.mm.1 | TC0200005130.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417464_at | - | - |
| Mouse Expression Set 430 | 1417464_PM_at | - | - |
| Murine Genome U74v2 Set | 162325_f_at | - | - |
| Murine Genome U74v2 Set | 162325_PM_f_at | - | - |
| Murine Genome U74v2 Set | 93259_at | - | - |
| Murine Genome U74v2 Set | 93259_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417464_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417464_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417464_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417464_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 755459 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6420484 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006420484 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2456978 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2456978 | - | - |
| mouse-6 v1.1 (Illumina) | scl18350.6.1_7-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6420484 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18350.6.1_7-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420484 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2882658 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2882658 | - | - |

Tnnc2
Genomic Map