Assay Details
| Gene Symbol: Csnk2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007788.3 | NP_031814.2 | 1 - 2 | 150 | 78 | |
| GenBank mRNA | AK011501.1 | - | 1 - 2 | 56 | 77 | |
| AK031617.1 | - | 1 - 2 | 115 | 78 | ||
| AK133980.1 | - | 1 - 2 | 78 | 78 | ||
| AK146032.1 | - | 1 - 2 | 109 | 78 | ||
| AK158077.1 | - | 1 - 2 | 80 | 78 | ||
| BC026149.1 | - | 1 - 2 | 54 | 4188101 | 78 | |
| BC060742.1 | - | 1 - 2 | 105 | 4945773 | 78 | |
| BY015187.1 | - | 1 - 2 | 150 | 78 | ||
| U51866.1 | - | 1 - 2 | 92 | 77 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the AK011501.1,AK031617.1,AK146032.1,AK158077.1,BC026149.1,BC060742.1,NM_007788.3,U51866.1 transcripts.
This assay is in the 5'UTR region of the AK011501.1,AK031617.1,AK146032.1,AK158077.1,BC026149.1,BC060742.1,NM_007788.3,U51866.1 transcripts.
Set Membership: |
|

Csnk2a1
Genomic Map