Assay Details
| Gene Symbol: Csnk2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007788.3 | NP_031814.2 | 3 - 4 | 474 | 125 | |
| GenBank mRNA | AK011501.1 | - | 3 - 4 | 380 | 125 | |
| AK031617.1 | - | 3 - 4 | 439 | 125 | ||
| AK146032.1 | - | 3 - 4 | 433 | 125 | ||
| AK158077.1 | - | 3 - 4 | 404 | 125 | ||
| BC026149.1 | - | 3 - 4 | 378 | 4188101 | 125 | |
| BC060742.1 | - | 3 - 4 | 429 | 4945773 | 125 | |
| BC089343.1 | - | 3 - 4 | 352 | 30360017 | 125 | |
| U17112.1 | - | 2 - 3 | 213 | 125 | ||
| U51866.1 | - | 3 - 4 | 416 | 125 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Csnk2a1
Genomic Map