Assay Details
| Gene Symbol: Fkbp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB241120.1 | - | 1 - 2 | 38 | 58 | |
| AB241121.1 | - | 1 - 2 | 38 | 58 | ||
| AB241122.1 | - | 1 - 2 | 38 | 58 | ||
| AF483488.1 | - | 1 - 2 | 38 | 58 | ||
| AF483489.1 | - | 1 - 2 | 38 | 58 | ||
| AK002777.1 | - | 1 - 2 | 145 | 58 | ||
| AK010693.1 | - | 1 - 2 | 169 | 58 | ||
| AK019362.1 | - | 1 - 2 | 173 | 58 | ||
| AK141261.1 | - | 1 - 2 | 153 | 58 | ||
| AK145400.1 | - | 1 - 2 | 162 | 58 | ||
| AK154751.1 | - | 1 - 2 | 117 | 58 | ||
| AK168333.1 | - | 1 - 2 | 153 | 58 | ||
| AK169186.1 | - | 1 - 2 | 76 | 58 | ||
| AK169242.1 | - | 1 - 2 | 160 | 58 | ||
| BC004671.1 | - | 1 - 2 | 120 | 3498682 | 58 | |
| BY065108.1 | - | 1 - 2 | 247 | 58 | ||
| BY108774.1 | - | 1 - 2 | 259 | 58 | ||
| CJ081656.1 | - | 1 - 2 | 263 | 58 | ||
| CX239711.1 | - | 1 - 2 | 125 | 58 | ||
| X60203.1 | - | 1 - 2 | 135 | 58 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Fkbp1a
Genomic Map