Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800009028.mm.1 |
NM_001035123.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009028.mm.1 |
NM_001035123.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009028.mm.1 |
TC0800001144.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009030.mm.1 |
NM_001035123.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009030.mm.1 |
NM_001035123.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009030.mm.1 |
TC0800001144.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009022.mm.1 |
NM_001035123.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009022.mm.1 |
NM_001035123.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009022.mm.1 |
TC0800001144.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009023.mm.1 |
NM_001035123.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009023.mm.1 |
NM_001035123.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009023.mm.1 |
TC0800001144.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009025.mm.1 |
NM_001035123.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009025.mm.1 |
NM_001035123.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009025.mm.1 |
TC0800001144.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009024.mm.1 |
NM_001035123.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009024.mm.1 |
NM_001035123.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009024.mm.1 |
TC0800001144.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009019.mm.1 |
TC0800001144.mm.1 |
MTA TC |