Assay Details
Gene Symbol: Ctcflos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK079954.1 | - | 177 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CCCTC-binding factor (zinc finger protein)-like | Ctcfl |
Boris, OTTMUSG00000016680 |
Anti-sense | Chr. 2 - 173093609 - 173119525 on NCBI Build GRCm38 | |
Downstream |
phosphoenolpyruvate carboxykinase 1, cytosolic |
Pck1 |
AI265463, PEPCK, Pck-1 |
23.8kb | Sense | Chr. 2 - 173153073 - 173159254 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
