Assay Details
| Gene Symbol: Ldhd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027570.3 | NP_081846.3 | 1 - 2 | 85 | 101 | |
| GenBank mRNA | AK014649.1 | - | 1 - 2 | 100 | 101 | |
| AK029820.1 | - | 99 | 101 | |||
| AY092768.1 | - | 1 - 2 | 90 | 101 | ||
| BC039155.1 | - | 1 - 2 | 81 | 4242682 | 101 | |
| BC055443.1 | - | 1 - 2 | 85 | 4235345 | 101 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014649.1, AY092768.1, BC039155.1, BC055443.1, NM_027570.3 transcripts.
Set Membership: |
|

Ldhd
Genomic Map