Assay Details
| Gene Symbol: St3gal2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009179.3 | NP_033205.2 | 5 - 6 | 1954 | 85 | |
| GenBank mRNA | AK029246.1 | - | 5 - 6 | 1869 | 85 | |
| AK031052.1 | - | 5 - 6 | 1897 | 85 | ||
| AK034863.1 | - | 5 - 6 | 1895 | 85 | ||
| AK053827.1 | - | 5 - 6 | 1893 | 85 | ||
| AK131653.1 | - | 4 - 5 | 777 | 85 | ||
| BC015264.1 | - | 4 - 5 | 1196 | 4211307 | 85 | |
| BC066064.1 | - | 5 - 6 | 1853 | 6850583 | 85 | |
| X76989.1 | - | 4 - 5 | 860 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

St3gal2
Genomic Map