Assay Details
Gene Symbol: Mlkl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029005.2 | NP_083281.1 | 3 - 4 | 585 | 74 | |
GenBank mRNA | AK018636.1 | - | 3 - 4 | 861 | 74 | |
AK170260.1 | - | 3 - 4 | 586 | 74 | ||
BC023755.1 | - | 3 - 4 | 870 | 5354568 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26167759 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26167760 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26167758 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046802 | AK170260.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046802 | BC023755.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046802 | NM_029005.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2046802 | AK018636.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 900761 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10876 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715873 | AF080623.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715873 | AF539811.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715873 | AK044891.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715873 | AK140337.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715873 | AK140347.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715873 | BC027669.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 715873 | NM_145831.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 514684 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 886373 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 129956 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122371 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 963330 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455592 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 158722 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254738 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71106 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 47267 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 929951 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 948766 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 704434 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579083 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 49357 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 27502 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18961 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108802 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123993 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145742 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046802 | AK170260.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046802 | BC023755.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046802 | NM_029005.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2046802 | AK018636.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108802 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145742 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123993 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06641899 | - | - |
Whole Mouse Genome G4122A | A_51_P440535 | AK170260.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P440535 | BC023755.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P440535 | NM_029005.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P440535 | AK018636.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110268 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105827 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108802 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145742 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123993 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06917686 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07355567 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07355569 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06917685 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07342527 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06917682 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26167758 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26167760 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26167759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167757 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167671 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167656 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167639 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167689 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167670 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167693 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167692 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167643 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167662 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167675 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167753 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167667 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167637 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167690 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167738 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167677 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167710 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167755 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167763 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167654 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167681 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167776 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167661 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167659 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167665 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167620 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167657 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167647 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167694 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167672 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167737 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167679 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26167712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07355567 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167780 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167766 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167779 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07299649 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167735 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167778 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167730 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167768 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167722 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06869895 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167765 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167723 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06690415 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167628 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167747 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167769 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167760 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167770 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167775 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167776 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167738 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167744 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167758 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07199975 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06917690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06763021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06982549 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167720 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167729 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167745 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06905978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167726 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167742 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07299645 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167737 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167740 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167781 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167772 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167753 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07355569 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06917683 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167751 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167761 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26167718 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10581818 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581822 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581820 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581815 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581819 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581821 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581823 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581814 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581816 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581817 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429570_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10581813 | - | - |
Mouse Genome 430 2.0 Array | 1429570_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1429570_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74568_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74568_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800022719.mm.1 | NM_029005.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800022719.mm.1 | TC0800002947.mm.1 | MTA TC |
Mouse Expression Set 430 | 1429570_at | - | - |
Mouse Expression Set 430 | 1429570_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429570_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429570_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2504397 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3006767 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3006767 | - | - |
