Assay Details
| Gene Symbol: Aars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146217.4 | NP_666329.2 | 3 - 4 | 776 | 67 | |
| GenBank mRNA | AK044451.1 | - | 3 - 4 | 527 | 67 | |
| AK085725.1 | - | 3 - 4 | 717 | 67 | ||
| AK150227.1 | - | 3 - 4 | 704 | 67 | ||
| AK158139.1 | - | 3 - 4 | 725 | 67 | ||
| AK193480.1 | - | 1 - 2 | 73 | 67 | ||
| AK207135.1 | - | 1 - 2 | 73 | 67 | ||
| AK207689.1 | - | 1 - 2 | 73 | 67 | ||
| AK208789.1 | - | 1 - 2 | 73 | 67 | ||
| AY223875.1 | - | 3 - 4 | 716 | 67 | ||
| BC033273.1 | - | 651 | 4196789 | 67 | ||
| BM943271.1 | - | 3 - 4 | 421 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aars
Genomic Map