Assay Details
| Gene Symbol: Aars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146217.4 | NP_666329.2 | 4 - 5 | 920 | 75 | |
| GenBank mRNA | AK036738.1 | - | 4 - 5 | 531 | 75 | |
| AK044451.1 | - | 4 - 5 | 671 | 75 | ||
| AK085725.1 | - | 4 - 5 | 861 | 75 | ||
| AK150227.1 | - | 4 - 5 | 848 | 75 | ||
| AK158139.1 | - | 4 - 5 | 869 | 75 | ||
| AK207135.1 | - | 2 - 3 | 217 | 75 | ||
| AY223875.1 | - | 4 - 5 | 860 | 75 | ||
| BC013261.1 | - | 1 - 2 | 145 | 75 | ||
| BC033273.1 | - | 795 | 4196789 | 75 | ||
| BM943271.1 | - | 4 - 5 | 565 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aars
Genomic Map