Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146216.2 | NP_666328.2 | 5 - 6 | 847 | 73 | |
| GenBank mRNA | AK031952.1 | - | 5 - 6 | 882 | 73 | |
| AK043319.1 | - | 5 - 6 | 879 | 73 | ||
| AK046826.1 | - | 5 - 6 | 819 | 73 | ||
| AK084520.1 | - | 5 - 6 | 898 | 73 | ||
| AK140191.1 | - | 5 - 6 | 874 | 73 | ||
| AK171577.1 | - | 5 - 6 | 887 | 73 | ||
| BC052199.1 | - | 5 - 6 | 847 | 6512678 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vac14
Genomic Map