Assay Details
| Gene Symbol: Phlpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122594.2 | NP_001116066.2 | 13 - 14 | 2100 | 61 | |
| GenBank mRNA | AK048260.1 | - | 13 - 14 | 2057 | 61 | |
| BC117961.1 | - | 13 - 14 | 2100 | 40098281 | 61 | |
| BC117962.1 | - | 13 - 14 | 2092 | 40098282 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phlpp2
Genomic Map