Assay Details
Gene Symbol: Tat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146214.3 | NP_666326.1 | 2 - 3 | 334 | 60 | |
GenBank mRNA | AK090244.1 | - | 2 - 3 | 332 | 60 | |
AK149383.1 | - | 2 - 3 | 332 | 60 | ||
AK149526.1 | - | 2 - 3 | 332 | 60 | ||
BC023949.1 | - | 2 - 3 | 318 | 5310059 | 60 | |
BC024120.1 | - | 2 - 3 | 309 | 5097591 | 60 | |
BC024264.1 | - | 1 - 2 | 257 | 5098682 | 60 | |
BC025934.1 | - | 2 - 3 | 269 | 5097113 | 60 | |
BC028821.1 | - | 2 - 3 | 309 | 5097589 | 60 | |
BC030728.1 | - | 2 - 3 | 313 | 5099036 | 60 | |
BC030729.1 | - | 2 - 3 | 307 | 5099570 | 60 | |
BC037526.1 | - | 2 - 3 | 289 | 5097795 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
