Assay Details
| Gene Symbol: Tepp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028532.3 | NP_082808.1 | 2 - 3 | 157 | 93 | |
| NM_199455.2 | NP_955534.1 | 3 - 4 | 382 | 93 | ||
| GenBank mRNA | AK006802.1 | - | 2 - 3 | 144 | 93 | |
| BC087870.1 | - | 3 - 4 | 378 | 6707505 | 93 | |
| BC104382.1 | - | 2 - 3 | 156 | 93 | ||
| BC104383.1 | - | 2 - 3 | 93 | |||
| BY706785.1 | - | 2 - 3 | 142 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK006802.1, BC104382.1, BC104383.1, NM_028532.3 transcripts.
Set Membership: |
|

Tepp
Genomic Map