Assay Details
| Gene Symbol: Tepp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028532.3 | NP_082808.1 | 4 - 5 | 391 | 112 | |
| NM_199455.2 | NP_955534.1 | 5 - 6 | 616 | 112 | ||
| GenBank mRNA | AK006802.1 | - | 4 - 5 | 378 | 112 | |
| BC087870.1 | - | 5 - 6 | 612 | 6707505 | 112 | |
| BC104382.1 | - | 4 - 5 | 390 | 112 | ||
| BC104383.1 | - | 4 - 5 | 391 | 112 | ||
| BY706785.1 | - | 4 - 5 | 376 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tepp
Genomic Map