Assay Details
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009408.2 | NP_033434.2 | 10 - 11 | 1187 | 157 | |
| GenBank mRNA | D10061.1 | - | 10 - 11 | 854 | 157 | |
| L20632.1 | - | 10 - 11 | 851 | 157 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Top1
Genomic Map