Assay Details
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009408.2 | NP_033434.2 | 11 - 12 | 1316 | 113 | |
| GenBank mRNA | CO425650.1 | - | 1 - 2 | 121 | 113 | |
| D10061.1 | - | 11 - 12 | 983 | 113 | ||
| L20632.1 | - | 11 - 12 | 980 | 113 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Top1
Genomic Map