Assay Details
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009408.2 | NP_033434.2 | 12 - 13 | 1495 | 91 | |
| GenBank mRNA | CO425650.1 | - | 2 - 3 | 300 | 91 | |
| D10061.1 | - | 12 - 13 | 1162 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Top1
Genomic Map