Assay Details
| Gene Symbol: Top1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009408.2 | NP_033434.2 | 15 - 16 | 1971 | 94 | |
| GenBank mRNA | CF734074.1 | - | 3 - 4 | 373 | 94 | |
| D10061.1 | - | 15 - 16 | 1638 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Top1
Genomic Map