Assay Details
| Gene Symbol: Narf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026272.3 | NP_080548.3 | 6 - 7 | 746 | 78 | |
| GenBank mRNA | AK013432.1 | - | 6 - 7 | 705 | 78 | |
| AK076232.1 | - | 6 - 7 | 729 | 78 | ||
| AK081305.1 | - | 6 - 7 | 730 | 78 | ||
| BC016090.1 | - | 6 - 7 | 685 | 4503713 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Narf
Genomic Map