Assay Details
| Gene Symbol: Hexdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001333.2 | NP_001001333.2 | 4 - 5 | 739 | 74 | |
| NM_001146073.1 | NP_001139545.1 | 3 - 4 | 325 | 74 | ||
| GenBank mRNA | AK154237.1 | - | 4 - 5 | 713 | 74 | |
| BC056487.1 | - | 4 - 5 | 2320 | 74 | ||
| BC069960.1 | - | 3 - 4 | 277 | 5007884 | 74 | |
| FM204885.1 | - | 3 - 4 | 276 | 74 | ||
| FM204886.1 | - | 3 - 4 | 276 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC069960.1 transcripts.
Set Membership: |
|

Hexdc
Genomic Map